VWR® Disposable Pellet Mixers and Cordless Motor
Supplier: VWR International
47747-358 47747-360 47747-362 47747-364 47747-366 47747-368 47747-370 47747-372
47747-358PK 259.3 CAD
47747-358 47747-360 47747-362 47747-364 47747-366 47747-368 47747-370 47747-372
VWR® Disposable Pellet Mixers and Cordless Motor
Homogenizers Tissue Grinders
Designed to homogenize cells and plant tissue or to re-suspend protein and DNA pellets.
  • Polybutylene terephthalate construction
  • DNase-, RNase-, and pyrogen-free
  • Grooved grip
  • Lightweight and ergonomically designed
Pestles and microtubes are manufactured from polybutylene terephthalate (PBTP) in an ISO Class 7 (FED-STD-209E Class 10,000/M5.5) cleanroom
Optional battery-powered motor can be used with pestles and microtubes. Motor is lightweight and greatly reduces hand fatigue. Its high-speed vortexing action completely homogenizes cell tissue andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable