Pleated (Fluted) Filter Paper, Grade 509, Ahlstrom
Supplier: Ahlstrom
5090-2400 5090-1250 5090-1500 5090-1850 5090-3200
28496-166PK 335.15 CAD
28496-166 47745-864 47745-866 47745-868 28334-192
Pleated (Fluted) Filter Paper, Grade 509, Ahlstrom
Filters Filter Papers Qualitative Filter Paper
Ahlstrom Grade 509 is the pre-pleated version of Grade 609.
Pleating improves the effective surface area which increases flow rate and loading capacity, thus decreasing filtration time.
Order Now
SPECIFICATIONS
Environmentally Preferable
Flow rate Medium
Load capacity Medium
Retention 4.0 µm