Axygen® Single- and Multi-Well Reservoirs, Corning
Supplier: Corning
RES-MW8-LP RES-MW12-LP RES-SW8-HP RES-SW12-HP RES-SW96-HP RES-SW1-LP RES-MW8-HP RES-MW12-HP RES-SW96-LP RES-MW12-HP-SI RES-SW12-HP-SI RES-SW1-LP-SI RES-SW384-HP-SI RES-SW384-LP-SI RES-SW96-LP-SI RES-SW8-HP-SI RES-MW12-LP-SI RES-MW4-HP RES-SW384-FX RES-SW384-FX-SI RES-MW8-LP-SI RES-SW384-LP RES-MW8-HP-SI RES-SW8-HP-S
12000-732CS 853.93 CAD
12000-732 12000-734 47743-954 47743-956 47743-958 47743-962 47743-966 47743-968 47744-808 47745-494 47745-498 47745-500 47745-502 47745-504 47745-508 47745-510 89179-654 89179-656 89179-658 89179-660 95042-900 47743-964 47745-496 CA10790-706
Axygen® Single- and Multi-Well Reservoirs, Corning
Reagent Reservoirs
These polypropylene reagent vessels are appropriate for automated and manual applications.
  • Reservoirs are made of clear virgin polypropylene for excellent chemical resistance
  • All reservoirs feature an SBS footprint for compatibility with most robotic systems
  • Flat-bottom reservoirs are single-well format and are guaranteed flat
Trough reservoirs incorporate flow troughs that hold the sample across the length or breadth of the reservoir in either 8- or 12-trough configurations. Diamond-bottom reservoirs feature 96- and 384-well profiles that are idealMore Product Information