Astral® Inoculation System, Bio Plas
Supplier: Bio Plas
7010 7000 7020
47742-594CS 796.02 CAD
47742-594 47742-592 47742-596
Astral® Inoculation System, Bio Plas
Spreaders Inoculating Loops and Needles
Disposable, color-coded system for quantitive procedures such as sampling, urine counts, serial dilutions, bacteriological inoculation, and streaking
The easy-to-use, rugged handle is designed for one-hand operation and will easily puncture the aluminum sealing film, attaching itself to either loop or needle. After the inoculation or streaking process is complete, used loops and needles can be ejected from the handle. The handle can be repeatedly autoclaved without damage.
Loops and needles are inserted into racks with 96 individual tubular chambers and are hermetically sealed under anMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable