Nitri-Pro™ Nitrile-Coated Gloves, Showa
Supplier: Showa
7199NCR-10
47742-276CS 1447.5 CAD
47742-276
Nitri-Pro™ Nitrile-Coated Gloves, Showa
Gloves
Tough, flexible nitrile (NBR) coating over a cotton jersey liner provides superior cut, puncture, snag, and abrasion resistance.
  • Sanitized
  • Hand-specific
  • Color: black
These gloves are ideal for handling rough, abrasive materials and have excellent resistance to oil and grease. They are launderable, making them an economical replacement for medium to heavy-duty leather palm work gloves. Choose the smooth finish for a good dry grip and more flexibility or the rough textured finish for a good wet grip and extra wear.
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Finish Textured
Cuff length 12.7 cm (5")