MicroSeal® 1200 Wipers, Berkshire
Supplier: Berkshire
MS12000909.8 MS12001212.6 MS1200404B10
21913-198CS 786 CAD
21913-198 21913-199 47732-718
MicroSeal® 1200 Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
MicroSeal® 1200 wipers are designed for spill control, equipment cleaning, and general cleanup in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments.
  • Critically low particles, fibers, ions, and extractables
  • High abrasion resistance
  • Chemically compatible with IPA, Acetone, and other solvents
  • Weight and caliper provide an excellent hand for cleaning rough surfaces
Continuous-filament, double-knitted, 100% polyester fabric is highly absorbent and tear-resistant. Wipers have clean, pliable, sealed edges.
Ordering information: Laundered and packagedMore Product Information