VWR® 12" Test Sieves, Half Height, All Stainless Steel
Supplier: VWR International
47730-274 47730-300 47730-306 47730-310 47730-314 47730-318 47730-330 47730-346 10804-776 10804-790 10811-550 10811-552 10811-554 10811-556 10811-558 10811-560 10811-562 10811-564 10811-566 10811-568 10811-570 10811-572 10811-574 10811-576 10811-578 10811-580 10811-582 10811-584 10811-586 10811-588 10811-634 10811-636 10811-638 10811-640 10811-642 10811-644 10811-646 10811-648 10811-650 10811-652 10811-654 10811-656 10811-658 10811-660 10811-662 10811-664
47730-274EA 724.1 CAD
47730-274 47730-300 47730-306 47730-310 47730-314 47730-318 47730-330 47730-346 10804-776 10804-790 10811-550 10811-552 10811-554 10811-556 10811-558 10811-560 10811-562 10811-564 10811-566 10811-568 10811-570 10811-572 10811-574 10811-576 10811-578 10811-580 10811-582 10811-584 10811-586 10811-588 10811-634 10811-636 10811-638 10811-640 10811-642 10811-644 10811-646 10811-648 10811-650 10811-652 10811-654 10811-656 10811-658 10811-660 10811-662 10811-664
VWR® 12" Test Sieves, Half Height, All Stainless Steel
Sieves Test Sieves
VWR® 12" diameter (304.8 mm ) all stainless steel test sieves are durable and ideal for sieving applications where a larger sample size is required.
  • Half Height (1⁵/₈") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Stainless steel construction
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying the sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes, and Centerline® sieve inspection or calibrationMore Product Information