VWR® 12" Test Sieves, Full Height, Brass and Stainless Steel
Supplier: VWR International
47730-156 47730-160 47730-162 47730-168 47730-170 47730-174 47730-180 47730-182 47730-184 47730-188 47730-190 47730-192 47730-194 47730-196 47730-198 47730-200 47730-202 47730-206 47730-208 47730-210 47730-212 47730-214 47730-218 47730-220 47730-222 47730-224 47730-226 47730-228 47730-230 47730-232 47730-234 47730-216 10811-536 10811-538 10811-540 10811-542 10811-546 10811-548
47730-156EA 524.86 CAD
47730-156 47730-160 47730-162 47730-168 47730-170 47730-174 47730-180 47730-182 47730-184 47730-188 47730-190 47730-192 47730-194 47730-196 47730-198 47730-200 47730-202 47730-206 47730-208 47730-210 47730-212 47730-214 47730-218 47730-220 47730-222 47730-224 47730-226 47730-228 47730-230 47730-232 47730-234 47730-216 10811-536 10811-538 10811-540 10811-542 10811-546 10811-548
VWR® 12" Test Sieves, Full Height, Brass and Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm) brass frame with stainless steel wire cloth test sieves are durable and ideal for sieving applications where a larger sample size is required.
  • Full Height (31/4") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Brass and stainless steel construction
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline®More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable