SP Bel-Art Scoops, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36748-0000 F36750-0000 F36747-0000 F36749-0000 F36753-0000 F36756-0000
47727-708CS 445.18 CAD
47727-708 47727-710 56920-020 56920-025 56920-036 56920-047
SP Bel-Art Scoops, Polypropylene, Bel-Art Products, a part of SP
Scoops
These scoops are rigid, strong, and smooth.
  • Flat-bottomed design
  • Autoclavable
  • Longer handle increases safe distance from sample
  • Polypropylene construction
A flat bottom keeps them upright when filled or empty, allowing them to be used as weighing boats or to be placed on a flat surface
Steam autoclavable at 121°C (250°F), they comply with FDA regulations for food contact.
The 60mL and 125mL scoops have a longer handle that increases the distance between the hand and sample.
Ordering information: Special prices on yearly contracts for largeMore Product Information