Clearview Classroom Stream Table
Supplier: Science First
651-6215 035590
470136-880EA 1251.39 CAD
470136-880 470358-450
Clearview Classroom Stream Table
Models Stream Models
An Essential Piece of Equipment for Any Earth Science Class
When it comes to studying erosion, sediment deposition, and the behavior of rivers and lakes, a stream table is a must-have. This affordable yet rugged ABS device features an integrated design that makes set-up a breeze – simply add water, turn on the pump, and teach! Additional features include five water jets – one water jet yields a little stream, while multiple jets form tributaries that join to form rivers, just like the natural landscape. A removable dam also creates a lake to illustrate how sediments form deltas. There’sMore Product Information