Wallcur® PRACTI-Code Meds
Supplier: WALLCUR, LLC.
1500CP 1501A 1502B 1503C 1504D 1505E 1506L 1507N 1508AR 1509AY 1510CP 1522LX 113HF 115TC TCCC-100 1514SF 1518L 1519B 1505AA
470232-714KT 67.79 CAD
470232-714 470232-716 470232-718 470232-720 470232-722 470232-724 470232-726 470232-728 470232-730 470232-732 470232-734 470232-742 470355-020 470355-030 470354-996 470354-990 470354-928 470354-986 470358-272
Wallcur® PRACTI-Code Meds
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Preloaded Syringes For Your EMS, ACLS Or Nursing Training
  • Contains No Medication
  • Luer Lock Syringes Labeled And Prefilled
  • Individually Packaged For Loading Into Practice Crash Carts Or Field Bags
Fill or Refill your training response kits with these commonly used code meds. Each syringe is preloaded to accurately simulate the experience of drug administration in a pressure simulation. The luer lock connection matches up perfectly with IV connections. Supplied in 5 ml, 10 ml or 50 ml syringes, each is individually packaged and labeled to simulate aMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable