Med Dimensions® Feline Intubation Trainer
Supplier: MED DIMENSIONS, LLC
MD-E-F1021E MD-E-F1021S MD-E-F1021S-1 MD-E-F1021S-2 MD-E-F1031S MD-E-F1031S-1 MD-E-F1041S
470357-264EA 302.4 CAD
470357-264 470357-260 470357-224 470357-172 470357-214 470357-284 470357-244
Med Dimensions® Feline Intubation Trainer
Educational Materials Biology Educational Materials Animal Biology Learning Activites
Ready to take your feline intubation skills to the next level?
  • Replicates a real feline larynx using real CT scans for precise training
  • Mimics the feel of a real larynx for accurate manipulation with fingers, tubes, or laryngoscope
  • Provides immediate visual and tactile feedback to refine intubation skills
  • Durable and reusable model withstands countless practice sessions without wearing down
The Feline Intubation Trainer offers a realistic platform for mastering critical airway management skills. Available in three configurations to accommodate diverseMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable