DC Digital Ammeter with Connectors
Supplier: GSC International
N-00002 N-00002-10
470357-010EA 86.9 CAD
470357-010 470357-024
DC Digital Ammeter with Connectors
Electrical Meters Ammeters
The 3A, 1mA Digital DC Ammeter with high accuracy is small and light enough to connect with any circuit.
  • Two alligator clips are directly connected with the meter’s main body for accurate measurements
  • A 3 volt lithium battery is included
The Digital DC Ammeter carries electric current even when powered off, providing quick and highly accurate measurements with one-push start-up. Provided with a zero-point adjustment function that ensures an accurate reading, the Digital DC Ammeter displays measured values digitally with plus or minus sign.