Ward's® Periodic Table Wall Charts
Supplier: Zenger Group
PTABLEBANNERBW PTABLEBANNERCOLOR 470357-314 470357-312
470356-540EA 293.88 CAD
470356-540 470356-542 470357-314 470357-312
Ward's® Periodic Table Wall Charts
Charts
Beautiful, up-to-date, and large enough for the whole class to see.
  • Updated names and atomic weights based on 2022 IUPAC published values
  • Download the matching student handout for free
  • Easy to read, even from across the room
  • Made to last: printed on heavy-duty, 13 mil vinyl, with grommets for easy and secure hanging
This giant periodic table wall chart is perfect for any Chemistry or Physical Science classroom. Measuring 48 inches tall, but only 62 inches wide, this full-sized wall chart fits large and smaller spaces alike. The beautiful font featuresMore Product Information