Archimedes' Principle Apparatus
Supplier: GSC International
ARCH-KT
470356-474EA 24.5 CAD
470356-474
Archimedes' Principle Apparatus
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Teach density the way Archimedes imagined it.
  • Each plastic cylinder is graduated, and measures 100 ml each
  • Perfect for developing the density concept using graphing
  • Instructions included
Archimedes' principle indicates that the upward buoyant force that is exerted on a body immersed in a fluid, whether fully or partially submerged, is equal to the weight of the fluid that the body displaces. The Archimedes' principle apparatus is perfect for attempting repeat experiments of this concept by using different volumes.
Accessories information: SpringMore Product Information