PicoSolutions Horizontal Wind Energy Plus Kit
Supplier: PICOSOLUTIONS GROUP INC SE
PS-3001-02 PS-3001-02-D1 PS-3001-02-D2 PS-3001-02-D3 PS-3001-02-D4 PS-3001-02-PK
470355-298EA 155.55 CAD
470355-298 470355-386 470355-338 470355-306 470355-392 470355-344
PicoSolutions Horizontal Wind Energy Plus Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Explore horizontal wind energy with a DIY hands-on turbine model, now including AC and DC power generation.
  • 3rd grade to college level tailored activities for different learning stages
  • Subjects spanning sustainable energy, math, aerodynamics, geometry
  • Easy 10 minute assembly
Build and test a wind turbine to teach the power of the wind Features a DC motor for beginners and an AC generator with 12+ experiments for advanced learners. The kit uses an LED, a buzzer, or a multimeter to show how wind generates electricity. The kit includes all parts forMore Product Information