Raspberry Pi 4 Computers
Supplier: NEWARK ELECTRONICS
02AH3164 03AH7034 29AK8617 64AH2041
470354-010EA 109 CAD
470354-010 470354-024 470354-020 470354-022
Raspberry Pi 4 Computers
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Ground-breaking increases in speed and performance compared to the Raspberry Pi 3 Model B+.
  • Choose either 4 or 8 GB of RAM
  • Single board computer only; power adapter, accessories, display cable, and microSD card available separately
For the end user, Raspberry Pi 4 Model B provides desktop performance comparable to entry-level x86 PC systems. This product’s key features include a high-performance 64-bit quad-core processor, dual-display support at resolutions up to 4K via a pair of micro-HDMI ports, hardware video decode at up to 4Kp60, 4 GB of RAM,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable