Test Tube Racks for 25 mm Tubes
Supplier: United Scientific Supplies
77804 77809
470353-646EA 29.09 CAD
470353-646 470353-632
Test Tube Racks for 25 mm Tubes
Racks Tube Racks
Polycarbonate racks provide clear views to tubes.
  • 3-tier design
  • Transparent plates for clear view of tubes
  • Holds tubes up to 25 mm in Ø
These polycarbonate racks with a 3-tier design provide clear view of the tube contents and convenient insertion and removal of tubes. Upper and middle plates are transparent. Beveled holes on bottom plate provide stability to standing tubes. Racks are autoclavable and can withstand sub-freezing temperatures. Available in 12-tube and 18-tube configurations, each able to hold tubes up to 25 mm in Ø.
Order Now
SPECIFICATIONS
Environmentally Preferable