HotGuard™ Autoclave Safety Gloves
Supplier: MTC BIO, INC.
A8095
470353-200PK 147.73 CAD
470353-200
HotGuard™ Autoclave Safety Gloves
Gloves
HotGuard™ Autoclave gloves are made of extra-thick cowhide leather for superior personal protection against hot glass and metal.
  • Heat resistant to 260 °C (500 °F)
  • Universal, one size fits all
  • 14" length provides wrist and forearm protection
  • 3-layer design offers superior heat resistance
A heat resistant cotton lining ensures the gloves are soft and comfortable to the touch, even during prolonged usage. Produced in a universal one size fits all design, HotGuard™ gloves are 14" in length to protect not only hands, but also wrists and forearms.