ACCU-SCOPE® EXS-210 Stereomicroscopes
Supplier: ACCU-SCOPE INC SE
EXS-210-123 EXS-210-124 EXS-210-13 EXS-210-24
470351-184EA 605.8 CAD
470351-184 470351-214 470351-202 470351-244
ACCU-SCOPE® EXS-210 Stereomicroscopes
Microscopes
Setting a new standard for quality, versatility and performance in a compact stereo microscope.
  • 1×, 2×, 3× and or 4× objective magnifications
  • Binocular viewing head
  • Inclined 45 degrees, rotatable 360 degrees
  • WF10×/20 mm eyepieces with eye guards
  • Frosted glass and black and white plates, dust cover
  • Variable reflected and transmitted rechargeable LED illumination
EXS-210 series offers educators and industry an exceptional value at an affordable price. An economical stereo microscope with superior optical performance, the EXS-210 stereo microscopeMore Product Information