ACCU-SCOPE® EXM-150 Monocular Microscopes
Supplier: ACCU-SCOPE INC SE
EXM-150 EXM-150-EP EXM-150-I EXM-150-IT EXM-150-MS EXM-150-T-WIFI EXM-150-WIFI EXM-151 EXM-151-DF EXM-151-EP EXM-151-T EXM-151-T-EP EXM-151-T-WIFI EXM-151-VT EXM-151-WIFI
470351-108EA 540.76 CAD
470351-108 470351-146 470351-226 470351-136 470351-182 470351-140 470351-158 470351-102 470351-094 470351-238 470351-080 470351-152 470351-070 470351-220 470351-256
ACCU-SCOPE® EXM-150 Monocular Microscopes
Microscopes
Monocular microscopes are easy to use and student friendly.
  • Monocular, dual inclined or inclined/vertical teaching head, rotatable 360°
  • DIN Achromat 4×, 10×, and 40×R* objectives; EXM-151 models also include a 100×R* oil immersion objective (*spring-loaded)
  • WF10× eyepieces, 18 mm field of view with pointer
  • Rear facing three-hole, ball-bearing nosepiece with positive click stops
  • Quadruple nosepiece is provided on the EXM-151 models
  • Separate coarse and fine focus adjustment controls
  • 119×119 mm stage with stage clips, or factory-installed double-plate
More Product Information