Celestron Nexyz DX Smartphone Adapter Kit
Supplier: Celestron International
81057
470348-474EA 160.85 CAD
470348-474
Celestron Nexyz DX Smartphone Adapter Kit
Telescopes
The NexYZ DX Kit is a great first step into the world of astroimaging or digiscoping.
  • A complete digiscoping solution
  • NexYZ connects a smartphone to an eyepiece while the Bluetooth shutter release button helps to achieve a steady shot every time
  • NexYZ’s patented design works with telescopes (with most 1.25" or 2" eyepieces), spotting scopes, binoculars, monoculars, and microscopes (with the included adapter rings)
  • Connect the remote shutter release to a phone via Bluetooth to take photos or start/stop video recording without unwanted vibration
  • Both devices
More Product Information