LED Lighted Rimless 2X Magnifier
Supplier: CARSON OPTICAL
RM-95
470347-526EA 27.8 CAD
470347-526
LED Lighted Rimless 2X Magnifier
Magnifiers
This 2X lighted 3.5" rimless acrylic round magnifier with soft pouch, is lightweight and ergonomically shaped.
  • Built-in LED
  • Distortion-free viewing
  • Includes batteries and soft pouch
Uses three, included, G10 batteries (389/LR1130).