SmartDrop™ X Spectrophotometer
Supplier: Ward's
NS1010 NS1010-E
470346-300EA 19385.99 CAD
470346-300 470346-312
SmartDrop™ X Spectrophotometer
Spectrophotometers and Photometers Biophotometers
The SmartDrop™ series of nano spectrophotometers provides quick and easy quantification of small samples.
  • Full-spectrum absorbance scans from 200 to 800 nm
  • Quickly calculates and displays 260/280 and 260/230 ratios
  • Long-life Xenon lamp with precision monochromator
  • Auto-ranging path lengths of 0.2 and 1.0 mm
  • Detect up to 4500 ng/µl of dsDNA without dilution
These spectrophotometers utilize a small sample volume which eliminates the need for dilutions, providing more accurate results. Each unit includes a full color, 7"
touchscreen with icon-drivenMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable