Eisco 6-Piece Plastic Graduated Beaker Set
Supplier: Wards
CH0137GSET
470345-554EA 43.89 CAD
470345-554
Eisco 6-Piece Plastic Graduated Beaker Set
Beakers
Perfect classroom starter set.
  • Set of 6 includes capacities of 50, 100, 250, 500, 1000 and 2000 ml
  • Made of premium quality polypropylene plastic
  • Superior chemical durability and heat tolerance
  • Excellent optical clarity
  • Raised graduations and specifications
  • Tapered spout for easy pouring
They are microwavable, autoclavable, and dishwasher safe for easy cleaning and sterilization. It can withstand temperatures between −20 to +135 °C. Beakers have raised graduation lines and feature a non-drip spout.