Glass Vacuum Desiccator with Stopcock
Supplier: Wards
CH0363C
470345-120EA 455.35 CAD
470345-120
Glass Vacuum Desiccator with Stopcock
Desiccators Desiccators, Glass
Borosilicate glass vacuum desiccator, fitted with a stopcock and porcelain desiccator plate, allowing controlled moisture and gas removal, ideal for laboratory experiments and sample storage.
  • Made from heavy-duty borosilicate 3.3 glass
  • Inner diameter of the chamber measures 24 cm (9.4")
  • Holds a full vacuum (740 mm Hg 20 cm) at room temperature for 24 hours - will remain airtight even if not under vacuum
  • Autoclavable, with superior chemical durability and heat tolerance
  • Low temperature gradient- can withstand high temperatures and thermal shocks
  • Designed
More Product Information