Eisco Glass Büchner Funnels with Plain Stem
Supplier: Wards
CH0466A CH0466B CH0466C CH0466D CH0466E
470344-636EA 21.8 CAD
470344-636 470345-078 470345-082 470345-658 470346-084
Eisco Glass Büchner Funnels with Plain Stem
Funnels Filtration Funnels Büchner Funnels
Büchner funnels with G3 porosity sintered discs made of borosilicate 3.3 glass.
  • Made of high-quality borosilicate 3.3 glass that is built to last and withstand multiple uses
  • Features a G3 porosity sintered disc
  • Straight, thick, uniform walls for increased durability, ease of use, and continual flow of liquid
Borosilicate glass Büchner funnels with straight sides and G3 porosity sintered discs. Eisco Labs Borosilicate glass has the following heat tolerances: Strain point 515 °C; Annealing point 565 °C; Softening point 820 °C.