Eisco Serological Pipettes, Class B

Supplier: Wards
CH0572F CH0572H CH0572G CH0572D
470344-300EA 9.59 CAD
470344-300 470345-058 470345-350 470345-950
Eisco Serological Pipettes, Class B
Pipettes Serological Pipettes
Class B color-coded Serological Pipettes made of high quality heavy duty borosilicate 3.3 glass with blue graduations.

  • Class B, conforms with DIN IS 835 standards
  • Calibrated for delivery to jet
  • Last drop expelled by blowing
  • Blue graduations for optimal contrast against clear liquids
  • Color coded for easy identification
  • Reusable

Class B tolerance, ±0.200 ml. Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.

Eisco Labs Borosilicate glass has the following heat tolerances:
-More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR