Eisco Rectangular Retort Bases, Cast Iron
Supplier: Wards
CH0653C CH0653B CH0653A CH0653D
470343-652EA 59.1 CAD
470343-652 470345-054 470345-116 470345-594
Eisco Rectangular Retort Bases, Cast Iron
Retort Stands
Heavy Retort Bases with tapped rod hole made of chemically-resistant coated cast iron.
  • Rectangular retort bases made of chemical-resistant cast iron with coated finished
  • Heavy and sturdy construction provides superb stability for experimental setups
  • Features a tapped hole for a 10×1.5 mm threaded rod (not included)
Heavy-weighted Retort Stands are made of chemically-resistant cast iron with a coated finish, and feature a tapped hole at one end, suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups.More Product Information