Wooden Pipette Rack
Supplier: Wards
CH0590B
470344-850EA 63.05 CAD
470344-850
Wooden Pipette Rack
Pipette Stands
Unpolished wood construction pipette rack with a capacity to hold 12 pipettes vertically, providing a cost-effective and functional storage option for pipettes.
  • Premium quality pipette stand featuring a sturdy wood construction that will stand up to the wear and tear of the lab
  • Rack holds pipettes vertically, allowing liquids to drain
  • Two legs extend backward to angle the rack slightly
  • Measures 12" wide and 14.75" tall, allowing it to fit in most lab spaces
  • Supports up to 12 pipettes
  • Designed for use in professional laboratories, but is an ideal choice for
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable