Eisco® High-Speed Low Friction Pulley on Rod
Supplier: Wards
PH0300SPL
470344-770EA 31.45 CAD
470344-770
Eisco® High-Speed Low Friction Pulley on Rod
Educational Materials Physics Educational Materials Mechanics Learning Activities
High-speed pulley with low friction design mounted on a rod for easy integration into experiments.
  • Pulley has a metal low friction bearing installed
  • Rod is made of durable aluminum; pulley is made of durable plastic
Pulley has a metal low friction bearing installed. Rod is made of durable aluminum and has a 12 mm diameter. Pulley is made of durable plastic.