Eisco® Metal Flywheel
Supplier: Wards
PH0313
470344-500EA 252.95 CAD
470344-500
Eisco® Metal Flywheel
Educational Materials Physics Educational Materials Mechanics Learning Activities
Balanced flywheel for exploring inertia and rotational dynamics.
  • Ball-bearing supported horizontal shaft
  • Bracket includes fixed pointer for observation
  • Can be wall-mounted using four integrated holes
Ideal for physics labs, this flywheel enables in-depth studies of rotational energy, kinetic energy, and inertia. The wheel is carefully balanced to ensure smooth motion and accurate results during demonstrations. While the apparatus is ready for use out of the box, additional weights can be introduced for extended experimentation (not included).
Order Now
SPECIFICATIONS
Environmentally Preferable