Eisco® Cast Iron Slotted Weight, 0.5 kg
Supplier: Wards
PH0264B1
470344-380EA 25.75 CAD
470344-380
Eisco® Cast Iron Slotted Weight, 0.5 kg
Weights Calibration Weights
Made of cast iron and painted black for durability and easy identification, this 0.5 kg slotted weight allows for precise measurements.
  • Ideal for use as spare/replacement weight
  • Painted black finish
  • 0.750" slot
  • 0.5 kg (1.10 pounds)
Eisco Labs cast iron slotted weight features an 0.750" slot and 3.625" Ext.Ø.