United Scientific™ DNA Model Kits with Four Models and CD
Supplier: United Scientific Supplies
DNAM01-K4 DNAM03-U
470339-732EA 382.45 CAD
470339-732 470339-982
United Scientific™ DNA Model Kits with Four Models and CD
Models Molecular Biology Models
Popular model for investigating DNA.
  • Classroom kit includes one preassembled DNA model, built to a scale of 10 cm to 1 nm, and four unassembled models
  • Central pillar support allows assembled model to rotate on circular stand
  • Each model can be used by a group of up to five students
  • Downloadable instructions explains key concepts related to DNA structure and includes step-by step instructions for model assembly
  • Contains small parts; not for children under three years
DNA model kit includes one preassembled DNA model, four unassembled models, andMore Product Information