Waterwise 4000 Countertop Distiller
Supplier: WATERWISE, INC. SE
4012 4506
470337-618EA 1287.65 CAD
470337-618 470337-620
Waterwise 4000 Countertop Distiller
Distillation Systems Automatic Distillation Systems
Compact, versatile water distillation at your fingertips.
  • Produces 1 gallon of 100% steam distilled water in 4 hours (up to 6 gpd)
  • Automatic shutoff after each 1-gallon cycle
  • Compact for storage and portability
With no installation or hookup needed, the Waterwise 4000 is the perfect water purification system for your classroom. Easy, convenient and portable, move it from class to class instantly for a steady stream of 100% pure steam distilled water. Simply fill the stainless steel boiler with tap water, press start and you're done.
Certifications:More Product Information