Substrate Specificity
Supplier: 3-D Molecular Designs
CSSSK-01
470337-150CS 1252.6 CAD
470337-150
Substrate Specificity
Educational Materials Biology Educational Materials AP Biology Learning Activities
Construct and examine a substrate
  • Interactive modeling
  • Build an enzyme from primary protein structure
  • Reusable
Help your students achieve that "aha moment" with this simple but elegant kit that demonstrates the highly-specific interaction between a substrate and an enzyme.
With the Substrate Specificity Kit© students can:
Use color-coded functional groups to construct a substrate and examine its chemical properties.
Use a mini toober to engineer an enzyme active site specific to the substrate constructed.
Explore different types of specificityMore Product Information