Making the Cut with CRISPR CAS-9
Supplier: 3-D Molecular Designs
CSMCC-01
470337-147CS 1236.8 CAD
470337-147
Making the Cut with CRISPR CAS-9
Educational Materials Biology Educational Materials AP Biology Learning Activities
How is CRISPR used in biotechnology?
  • Interactive modeling
  • How does CAS-9 'Make The Cut'
  • Reusable
Model how the CAS-9 protein is programmed with RNA to search for and identify specific sequences of DNA before making the double-stranded cut. Explore and discuss how CAS-9 is being engineered with new features that make it a useful tool for editing the human genome.
With making the cut with CRISPR-CAS9's colorful foam pieces and placemat your students can:
Build a schematic model of the CRISPR-CAS-9 RNA-guided endonuclease, including the viral double-More Product Information