biOrb® Heat And Light Accessories
Supplier: GEOGLOBAL PARTNERS
45588 45992 45997 45994 45995 72448 72461 72169 72168 73180 70371 73114
470234-738CS 408.8 CAD
470234-738 470234-842 470234-848 470234-886 470234-845 470234-847 470336-858 470336-924 470336-926 470336-928 470336-930 470336-932 470336-934
biOrb® Heat And Light Accessories
Animal Supplies Animal Environmental Enrichment
Improve Your biOrb® Aquaria.
  • Upgrade your older models
  • Expand the types of fish you can keep
A wider selection of fish can be kept by converting the aquarium to tropical. To do this the water needs to be warmed by an aquarium heater. The biOrb® Heater Pack contains a 50 W standard aquarium heater capable of heating the water in any biOrb® up to 16 gallons/60 L. A Heater Stand is also supplied that enables the heater to be fitted securely onto the curved surface of the biOrb®. It is also important to monitor the temperature of the aquarium with aMore Product Information