Klinger Light Box Multi Pack, 3 Colors
Supplier: Klinger
LTB-RD-KL LTB-BL-KL LTB-GR-KL LTB-MLT-KL
470336-002EA 60.99 CAD
470336-002 470336-004 470336-006 470336-008
Klinger Light Box Multi Pack, 3 Colors
Lights Light Boxes
Beautiful, bright and captivating classroom safe LED's in primary colors of light (red, green and blue) for color mixing, refraction and reflection.
  • Bring in additive color with one fun and inexpensive set of light box LED's
  • With the light boxes attached to whiteboard it doubles as a ray box
  • Research, examine and detect all the properties of light with one uncomplicated tool
  • There is no need for expensive pieces of science apparatus to make the science of light and optics fun and available for ages 7 to 18 and higher
  • A hands-on science tool for the
More Product Information