Economy Lab Tables and Replacement Tops
Supplier: Ward's Science
80A.1100 80B.1100 80C.1100 80D.1100 80A.1200 80B.1200 80C.1200 80D.1200 80A.1110 80B.1110 80C.1110 80D.1110 80A.1210 80B.1210 80C.1210 80D.1210 84A.1100 84B.1100 84C.1100 84D.1100 84A.1200 84B.1200 84C.1200 84D.1200 84A.1110 84B.1110 84C.1110 84D.1110 84A.1210 84B.1210 84C.1210 84D.1210 80A.1000 80B.1000 80C.1000 80D.1000 84A.1000 84B.1000 84C.1000 84D.1000
470335-100EA 921.15 CAD
470335-100 470335-102 470335-104 470335-106 470335-108 470335-110 470335-112 470335-114 470335-116 470335-118 470335-120 470335-122 470335-124 470335-126 470335-128 470335-130 470335-132 470335-134 470335-136 470335-138 470335-142 470335-144 470335-146 470335-148 470335-152 470335-154 470335-156 470335-158 470335-162 470335-164 470335-166 470335-168 470335-172 470335-174 470335-176 470335-178 470335-180 470335-182 470335-184 470335-186
Economy Lab Tables and Replacement Tops
Furniture Tables and Workbenches
Provide students a durable lab work surface with these economical tables.
  • Quick and easy assembly
  • Solid oak aprons and legs
  • Available with two different work surfaces: Thermofused laminate or Trespa TopLab Plus top
Tops are available as thermofused laminate, which is produced under heat and pressure to form a permanent thermal bond between the melamine laminate and a 1" high density substrate or as Trespa TopLab Plus tops, which are manufactured with an innovative technology that bonds chemical resistant resins and fibers into a solid ³/₄" compositeMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable