Ward's® HiPette Micropipettes
Supplier: DLAB SCIENTIFIC INC.
7030511004 7030511008 7030511014 7030511001 7030511005 7030511009 7030511040
470335-064EA 333.45 CAD
470335-064 470335-066 470335-068 470335-070 470335-072 470335-074 470335-076
Ward's® HiPette Micropipettes
Pipettes Micropipettes
HiPette is a line of high-performing science education micropipettes for when laboratory-grade precision is needed.
  • Has a 4-digit counter visible display and a counter increment of 1000th of the range
  • Filter element prevents volatile liquids from entry into the pipette and helps avoid liquid back-flow
  • Requires no calibration for up to 50000 pipetting operations
  • Autoclavable at 121 degrees Celsius
  • UV resistant and tolerant to strong chemical corrosion
The HiPette range offers an ergonomic and ambidextrous design for all day pipetting. It features aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable