Acrylic Color Filters, United Scientific
Supplier: United Scientific Supplies
COLORF-5
470332-748PK 7.45 CAD
470332-748
Acrylic Color Filters, United Scientific
Educational Materials Physics Educational Materials Optics Learning Activities
Set of 5 acrylic filters.
  • Filters measure 2.5×2.5×0.125"
  • Includes one each of red, orange, yellow, blue, and green
Set of five color filters made of acrylic. Includes one each of the following colors: red, orange, yellow, blue, and green.