Eisco LabGlass® Distilling Flasks with Oblique Neck, Round Bottom, Interchangable Joints
Supplier: Wards
CH0419A CH0419B CH0419C CH0419D CH0419E CH0419F CH0419G CH0419H
470332-146EA 26.4 CAD
470332-146 470332-148 470332-150 470332-152 470332-154 470332-156 470332-158 470332-160
Eisco LabGlass® Distilling Flasks with Oblique Neck, Round Bottom, Interchangable Joints
Flasks Distillation Flasks
High quality round bottom distilling flasks with oblique neck.
  • Made of high quality borosilicate glass 3.3
  • Interchangable, ground joints
  • Oblique neck design
  • Round bottom
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable