Eisco LabGlass® Distilling Flasks with 3 Parallel Necks, Round Bottom, Interchangable Joints
Supplier: Wards
CH0418A CH0418B CH0418C CH0418D CH0418E CH0418F CH0418G CH0418H CH0418I CH0418J
470332-102EA 48.2 CAD
470332-102 470332-104 470332-106 470332-108 470332-110 470332-112 470332-114 470332-116 470332-118 470332-120
Eisco LabGlass® Distilling Flasks with 3 Parallel Necks, Round Bottom, Interchangable Joints
Flasks Distillation Flasks
High quality round bottom distilling flasks with three parallel necks.
  • Made of borosilicate glass 3.3
  • Ground interchangable joint for secure fit with other glassware
  • Graduated
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C.
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed byMore Product Information