Sickle Cell Gene Detection (DNA-Based)
Supplier: Edvotek
116-C
470330-670EA 464.2 CAD
470330-670
Sickle Cell Gene Detection (DNA-Based)
Educational Materials Biology Educational Materials Genetics Learning Activities
Hands-on simulation of Sickle Cell diagnostic test.
  • Perform a simulated test on a patient sample
  • No actual blood or blood products are used.
  • Includes all required reagents
  • Use DNA Electrophoresis to reach a diagnosis
  • Activity can be completed in 45 minutes
Sickle Cell Anemia is a common genetic disease that causes long rods in red blood cells, giving them a 'sickled' appearance. These cells get stuck in small capillaries of the blood stream leading to oxygen deprivation that causes pain and organ damage.
Sickle Cell Anemia is caused by a singleMore Product Information