Innovating Science® Acids and Bases Lab Activity
Supplier: Ward's Science
IS1001 IS1500
470163-306KT 61.15 CAD
470163-306 470328-340
Innovating Science® Acids and Bases Lab Activity
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
In this activity, students make hypotheses regarding household items and then test them.
  • Perfect Introduction to Acids and Bases
  • Students Test Household Items
  • Kit Features Extra Litmus Paper for More Experiments
  • All Required Chemicals are Included
  • Acid and Base Chemistry in Microscale
Students learn about acids and bases through a class discussion and then test a dilute acid and dilute base with neutral litmus paper to learn how the paper indicates something is acidic or basic.
Once students master that skill, they can move on to test some commonMore Product Information