Glass Equilateral Prisms
Supplier: Wards
PH0554GG PH0554HG PH0554IG PH0554LG
470328-138EA 7.1 CAD
470328-138 470328-140 470328-142 470328-144
Glass Equilateral Prisms
Educational Materials Physics Educational Materials Optics Learning Activities
Designed for educational purposes and science classroom demonstrations in optics, light refraction, spectrometry, and wavelengths.
  • Made of optical quality glass
  • Polished on all sides
These glass prisms will create a nice rainbow (even in room light) by using the simple LED light of a smart phone. Users will need to create a small slit in a black piece of paper and use that slit in front of the LED, which will create a light ray. The prisms will then refract the white light and create a nice rainbow on a target away from the prism. The farther away youMore Product Information