Acrylic Equilateral Prisms

Supplier: Wards
PH0554AC PH0554BC PH0554CC PH0554DC PH0554SET
470328-130EA 4.9 CAD
470328-130 470328-132 470328-134 470328-136 470328-146
Acrylic Equilateral Prisms
Educational Materials Physics Educational Materials Optics Learning Activities
Designed for educational purposes and science classroom demonstrations of refraction principles and light physics.

  • Made of high quality acrylic
  • Refractive index almost identical to optical glass
  • Polished on all sides

High quality equilateral prisms are chip- and shatter-resistant, while having a refractive index very close to optical glass. They can produce a rainbow from a non-monochromatic light source such as sunlight. Acrylic is a softer material, so handle with care to prevent scratching.

Ordering information: The Acrylic Equilateral Prism KitMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR