Eisco® Safety Pack Glassware
Supplier: Wards
CH196031 CH196032 CH196033 CH196034 CH196041 CH196042 CH196046 CH196047 CH196048 CH196061
470328-044KT 37.75 CAD
470328-044 470328-046 470328-048 470328-050 470328-052 470328-054 470328-056 470328-058 470328-060 470328-062
Eisco® Safety Pack Glassware
Labware Sets
Convenient sets of glassware expertly packaged custom-fit foam inserts for worry-free transport and safe storage.
  • Convenient sets of high-quality Borosilicate 3.3 glassware
  • Packaged in thick, custom-fit foam inserts for safe transport and storage
  • Superior heat tolerance and chemical resistance
  • Autoclavable at 121 °C
  • Volumetric glassware is hand calibrated in Eisco's ISO 17025 compliant laboratory
Eisco safety pack glassware are all made of high-quality Borosilicate 3.3 glass. You will find these convenient sets of glassware expertly packaged in heavyMore Product Information