LabGlass Eisco™ Heavy-Duty Glass Beakers, Eisco Scientific LLC
Supplier: Wards
CH200000 CH200000PK12 CH200001 CH200001PK12 CH200002 CH200002PK12 CH200003 CH200003PK6 CH200004 CH200004PK6
470328-024EA 5.05 CAD
470328-024 470328-026 470328-028 470328-030 470328-032 470328-034 470328-036 470328-038 470328-040 470328-042
LabGlass Eisco™ Heavy-Duty Glass Beakers, Eisco Scientific LLC
Beakers
High quality Borosilicate 3.3 glass with 5 mm thick walls provide maximum durability and strength against breakage.
  • Uniform, thick walls provide increased durability and stability
  • Beaded rim and spout for easy pouring
  • Easy to read, chemical resistant white printed graduations
  • Superior heat tolerance and chemical resistance
  • Autoclavable at 121 °C
Eisco heavy duty beakers are made of high-quality Borosilicate 3.3 glass. These durable glass beakers have a low temperature gradient and can withstand high temperatures as well as thermal shocks. They areMore Product Information